COMH93D60E18 Combi-Blocks, Inc.

2-Amino-3-(3,4,5-trifluorophenyl)propanoic acid hydrochloride

  • MDL number MFCD29038823
  • Cas Number : 870483-31-1
  • Molecular Weight : 255.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9346
Availability details for
Quantity
  • Availability
  • Add to Cart