COMH93D60E24 Combi-Blocks, Inc.

1-(2,6-Dichloro-4-(perfluoropropan-2-yl)phenyl)-1h-pyrazole

  • MDL number MFCD30471865
  • Cas Number : 2007916-01-8
  • Molecular Weight : 381.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9360
Availability details for
Quantity
  • Availability
  • Add to Cart