COMH93D60E2A Combi-Blocks, Inc.

(E)-1-Methyl-3-(2-nitrovinyl)-1h-pyrrolo[2,3-b]pyridine

  • MDL number MFCD22121412
  • Cas Number : 1094709-34-8
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9368
Availability details for
Quantity
  • Availability
  • Add to Cart