COMH93D60E3C Combi-Blocks, Inc.

(2R)-2-Amino-2-(6-methoxy(3-pyridyl))ethan-1-ol hcl

  • MDL number MFCD24428874
  • Cas Number : 1641401-33-3
  • Molecular Weight : 204.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9391
Availability details for
Quantity
  • Availability
  • Add to Cart