COMH93D60E55 Combi-Blocks, Inc.

5-((2,3-Trans)-5-bromo-2-(4-hydroxyphenyl)-2,3-dihydrobenzofuran-3-yl)benzene-1,3-diol

  • MDL number MFCD29059410
  • Cas Number : 1632406-61-1
  • Molecular Weight : 399.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9425
Availability details for
Quantity
  • Availability
  • Add to Cart