COMH93D60E6D Combi-Blocks, Inc.

Docetaxel trihydrate

  • MDL number MFCD26960948
  • Cas Number : 1279037-32-9
  • Molecular Weight : 861.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9455
Availability details for
Quantity
  • Availability
  • Add to Cart