COMH93D60E70 Combi-Blocks, Inc.

N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-benzamide

  • MDL number MFCD22494317
  • Cas Number : 1330596-14-9
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9458
Availability details for
Quantity
  • Availability
  • Add to Cart