COMH93D60E7D Combi-Blocks, Inc.

3-Iodo-5-methyl-6-nitro-indazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31654484
  • Cas Number : 586330-46-3
  • Molecular Weight : 403.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9476
Availability details for
Quantity
  • Availability
  • Add to Cart