COMH93D60EAF Combi-Blocks, Inc.

5-Isobutoxy-isophthalic acid monomethyl ester

  • MDL number MFCD24039312
  • Cas Number : 1282520-84-6
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9536

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9536
Availability details for
Quantity
  • Availability
  • Add to Cart