COMH93D60ED0 Combi-Blocks, Inc.

2-[(6-Fluoropyridin-3-yl)oxy]acetonitrile

  • MDL number MFCD24190885
  • Cas Number : 1545052-36-5
  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9574
Availability details for
Quantity
  • Availability
  • Add to Cart