COMH93D60EF6 Combi-Blocks, Inc.

1-(4-Bromo-3,5-dimethylphenyl)-5,5-dimethylimidazolidine-2,4-dione

  • MDL number MFCD30803364
  • Cas Number : 1613373-40-2
  • Molecular Weight : 311.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9619
Availability details for
Quantity
  • Availability
  • Add to Cart