COMH93D60EFB Combi-Blocks, Inc.

T-Boc-n-amido-peg6-ch2co2h

  • MDL number MFCD30489670
  • Cas Number : 391684-36-9
  • Molecular Weight : 439.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9626
Availability details for
Quantity
  • Availability
  • Add to Cart