COMH93D60F12 Combi-Blocks, Inc.

3,6-Dibromo-9-heptyl-9h-carbazole

  • MDL number MFCD31618132
  • Cas Number : 1103535-99-4
  • Molecular Weight : 423.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9658
Availability details for
Quantity
  • Availability
  • Add to Cart