COMH93D60F73 Combi-Blocks, Inc.

7-Bromo-1,5-naphthyridin-4-amine

  • MDL number MFCD22571058
  • Cas Number : 1309774-12-6
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9778

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9778
Availability details for
Quantity
  • Availability
  • Add to Cart