COMH93D60F9E Combi-Blocks, Inc.

2-tert-Butyl 5-ethyl 2,7-diazaspiro[3.5]nonane-2,5-dicarboxylate

  • MDL number MFCD31555273
  • Cas Number : 2102410-34-2
  • Molecular Weight : 298.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9827
Availability details for
Quantity
  • Availability
  • Add to Cart