COMH93D60FC7 Combi-Blocks, Inc.

3-Methylimidazo[1,2-a]pyridine-5-carbaldehyde

  • MDL number MFCD22551989
  • Cas Number : 1315359-91-1
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9878

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9878
Availability details for
Quantity
  • Availability
  • Add to Cart