COMH93D610E5 Combi-Blocks, Inc.

Cyclopropylmethyl-(2-hydroxy-ethyl)-carbamic acid benzyl ester

  • MDL number MFCD31700312
  • Cas Number : 2142656-96-8
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0250
Availability details for
Quantity
  • Availability
  • Add to Cart