COMH93D610FD Combi-Blocks, Inc.

2-Bromo-n,6-dimethylbenzamide

  • MDL number MFCD31735324
  • Cas Number : 1856138-98-1
  • Molecular Weight : 228.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0279

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0279
Availability details for
Quantity
  • Availability
  • Add to Cart