COMH93D61123 Combi-Blocks, Inc.

5-Bromo-3-(cyclopropylmethyl)-1,3-benzoxazol-2(3h)-one

  • MDL number MFCD31725207
  • Cas Number : 1802149-42-3
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0328

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0328
Availability details for
Quantity
  • Availability
  • Add to Cart