COMH93D6114E Combi-Blocks, Inc.

3-Bromo-n-(2-fluoroethyl)benzamide

  • MDL number MFCD31811328
  • Cas Number : 1862836-07-4
  • Molecular Weight : 246.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0378
Availability details for
Quantity
  • Availability
  • Add to Cart