COMH93D61156 Combi-Blocks, Inc.

3-[2-(3-Bromo-phenyl)-ethoxy]-4-methoxy-benzoic acid methyl ester

  • MDL number MFCD32067028
  • Molecular Weight : 365.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0392
Availability details for
Quantity
  • Availability
  • Add to Cart