COMH93D61174 Combi-Blocks, Inc.

1-Benzyl 2-methyl cis-5-aminopiperidine-1,2-dicarboxylate hydrochloride

  • MDL number MFCD31925237
  • Molecular Weight : 328.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0427
Availability details for
Quantity
  • Availability
  • Add to Cart