COMH93D61183 Combi-Blocks, Inc.

4,5,6,7-Tetrahydro-1h-imidazol[4,5-c]-pyridine dihydrochloride hydrate

  • MDL number MFCD16875645
  • Molecular Weight : 214.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0445
Availability details for
Quantity
  • Availability
  • Add to Cart