COMH93D61184 Combi-Blocks, Inc.

N'-Hydroxy-2-[[3-(trifluoromethyl)pyridin-2-yl]thio]ethanimidamide

  • MDL number MFCD00208090
  • Cas Number : 263762-04-5
  • Molecular Weight : 251.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0446
Availability details for
Quantity
  • Availability
  • Add to Cart