COMH93D61237 Combi-Blocks, Inc.

tert-Butyl 2-fluoro-4-(trifluoromethyl)benzoate

  • MDL number MFCD32176680
  • Molecular Weight : 264.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0657
Availability details for
Quantity
  • Availability
  • Add to Cart