COMH93D612CF Combi-Blocks, Inc.

Hex-5-enyl-methyl-carbamic acid 9h-fluoren-9-ylmethyl ester

  • MDL number MFCD32176826
  • Molecular Weight : 335.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0845
Availability details for
Quantity
  • Availability
  • Add to Cart