COMH93D6130E Combi-Blocks, Inc.

3-Bromo-n-cyclopropyl-4-(trifluoromethyl)benzamide

  • MDL number MFCD32176690
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0917
Availability details for
Quantity
  • Availability
  • Add to Cart