COMH93D6130F Combi-Blocks, Inc.

2-(4-Fluorophenyl)ethylamine hydrobromide

  • MDL number MFCD32062908
  • Cas Number : 1807536-06-6
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0918
Availability details for
Quantity
  • Availability
  • Add to Cart