COMH93D61339 Combi-Blocks, Inc.

4-Hex-1-ynylbenzaldehyde

  • MDL number MFCD00276593
  • Cas Number : 175203-90-4
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-0972
Availability details for
Quantity
  • Availability
  • Add to Cart