COMH93D6144C Combi-Blocks, Inc.

Methyl (r)-2-(2,3,4-trichlorophenoxy)butanoate

  • MDL number MFCD32196838
  • Molecular Weight : 297.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1301
Availability details for
Quantity
  • Availability
  • Add to Cart