COMH93D6156A Combi-Blocks, Inc.

4-(4,4-Difluoro-piperidin-1-ylmethyl)-2-isopropoxy-phenylamine

  • MDL number MFCD31700363
  • Cas Number : 2270911-25-4
  • Molecular Weight : 284.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1643
Availability details for
Quantity
  • Availability
  • Add to Cart