COMH93D61570 Combi-Blocks, Inc.

4-(3-Fluorophenyl)-3-nitropyridin-2-amine

  • MDL number MFCD32067029
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1650
Availability details for
Quantity
  • Availability
  • Add to Cart