COMH93D61572 Combi-Blocks, Inc.

8-(tert-Butyl-diphenyl-silanyloxy)-octan-1-ol

  • MDL number MFCD31735367
  • Cas Number : 94124-45-5
  • Molecular Weight : 384.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1653
Availability details for
Quantity
  • Availability
  • Add to Cart