COMH93D615A8 Combi-Blocks, Inc.

7-Aminopyrido[4,3-d][1,2,4]triazolo[1,5-a]pyrimidin-8(7h)-one

  • MDL number MFCD31794361
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1721
Availability details for
Quantity
  • Availability
  • Add to Cart