COMH93D615D2 Combi-Blocks, Inc.

1-Bromo-3-fluoro-2-isopropoxy-5-nitro-benzene

  • MDL number MFCD31760357
  • Cas Number : 2301067-86-5
  • Molecular Weight : 278.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1770
Availability details for
Quantity
  • Availability
  • Add to Cart