COMH93D61636 Combi-Blocks, Inc.

(S)-1-tert-Butyl 3-(chloromethyl) piperidine-1,3-dicarboxylate

  • MDL number MFCD32067177
  • Molecular Weight : 277.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1883
Availability details for
Quantity
  • Availability
  • Add to Cart