COMH93D61652 Combi-Blocks, Inc.

1-(5-Bromo-7-methoxy-1-benzofuran-2-yl)ethan-1-one

  • MDL number MFCD02102416
  • Cas Number : 150612-66-1
  • Molecular Weight : 269.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1914
Availability details for
Quantity
  • Availability
  • Add to Cart