COMH93D6167F Combi-Blocks, Inc.

T-Boc-aminooxy-peg5-amine

  • MDL number MFCD31811543
  • Cas Number : 2250216-94-3
  • Molecular Weight : 396.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-1971
Availability details for
Quantity
  • Availability
  • Add to Cart