COMH93D6169E Combi-Blocks, Inc.

(3-Bromo-1h-pyrazol-5-yl)methanediol

  • MDL number MFCD31695981
  • Molecular Weight : 193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2009
Availability details for
Quantity
  • Availability
  • Add to Cart