COMH93D616B1 Combi-Blocks, Inc.

(5-Chloro-thiophen-2-yl)-(4-fluoro-piperidin-1-yl)-methanone

  • MDL number MFCD31725269
  • Cas Number : 2270910-81-9
  • Molecular Weight : 247.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2031
Availability details for
Quantity
  • Availability
  • Add to Cart