COMH93D616B6 Combi-Blocks, Inc.

Ethyl 3-((1r,4r)-4-((tert-butoxycarbonyl)amino)cyclohexyl)-3-oxopropanoate

  • MDL number MFCD31916185
  • Cas Number : 2340293-37-8
  • Molecular Weight : 313.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2036
Availability details for
Quantity
  • Availability
  • Add to Cart