COMH93D61715 Combi-Blocks, Inc.

2-(4-Fluoro-2-trifluoromethyl-phenoxy)-propionic acid

  • MDL number MFCD31760417
  • Cas Number : 2301068-72-2
  • Molecular Weight : 252.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2150
Availability details for
Quantity
  • Availability
  • Add to Cart