COMH93D61739 Combi-Blocks, Inc.

6-Chloro-2-iodo-3-propoxypyridine

  • MDL number MFCD31725263
  • Cas Number : 1865523-70-1
  • Molecular Weight : 297.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2195
Availability details for
Quantity
  • Availability
  • Add to Cart