COMH93D6175F Combi-Blocks, Inc.

2-((((9H-Fluoren-9-yl)methoxy)carbonyl)((1-(tert-butoxycarbonyl)piperidin-4-yl)methyl)amino)acetic acid

  • MDL number MFCD31803904
  • Cas Number : 2102408-73-9
  • Molecular Weight : 494.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2239
Availability details for
Quantity
  • Availability
  • Add to Cart