COMH93D61812 Combi-Blocks, Inc.

(Z)-Hex-3-en-1-yltriphenylphosphonium iodide

  • MDL number MFCD32196757
  • Cas Number : 21676-06-2
  • Molecular Weight : 472.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2461

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2461
Availability details for
Quantity
  • Availability
  • Add to Cart