COMH93D6193D Combi-Blocks, Inc.

Mal-peg5-t-butyl ester

  • MDL number MFCD31811537
  • Cas Number : 2250216-91-0
  • Molecular Weight : 445.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2827
Availability details for
Quantity
  • Availability
  • Add to Cart