COMH93D61952 Combi-Blocks, Inc.

(5-Bromo-2-iodo-benzyl)-cyclopropyl-amine

  • MDL number MFCD31760269
  • Cas Number : 1699080-49-3
  • Molecular Weight : 352.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2853
Availability details for
Quantity
  • Availability
  • Add to Cart