COMH93D6195D Combi-Blocks, Inc.

(2R,5S)-1,2,5-Trimethylpiperazine 2hcl

  • MDL number MFCD31807392
  • Cas Number : 2225878-97-5
  • Molecular Weight : 200.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-2867
Availability details for
Quantity
  • Availability
  • Add to Cart