COMH93D619DD Combi-Blocks, Inc.

2-(3-Bromo-4-methylphenoxy)acetonitrile

  • MDL number MFCD31725283
  • Cas Number : 1861576-55-7
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3030
Availability details for
Quantity
  • Availability
  • Add to Cart