COMH93D61A83 Combi-Blocks, Inc.

(4As,7ar)-1-tert-butyl 4a-ethyl 6-benzyl-4-oxooctahydro-1h-pyrrolo[3,4-b]pyridine-1,4a-dicarboxylate

  • MDL number MFCD31803778
  • Cas Number : 2177264-96-7
  • Molecular Weight : 402.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3220

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3220
Availability details for
Quantity
  • Availability
  • Add to Cart